Learn More
Medchemexpress LLC m-PEG12-azide | 2170098-29-8 | MFCD13184959 | 98.6% | C25H51N3O12 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1408301G
Encompass_Preferred
m-PEG12-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PROTACs degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
Catalog No. 50-003-53137