Learn More
Medchemexpress LLC M-PEG12-Br | 1620461-89-3 | C25H51BrO12 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1413781G
Encompass_Preferred
m-PEG12-Br is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules are designed to exploit the intracellular ubiquitin-proteasome system, selectively degrading target proteins. PROTACs feature two distinct ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets a specific protein, making m-PEG12-Br a key component in this targeted protein degradation strategy.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Connects ligands for E3 ubiquitin ligase and target protein
Catalog No. 50-003-55338