Learn More
Medchemexpress LLC M-PEG12-DBCO | 96.9% | C44H66N2O14 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140317100MG
Encompass_Preferred
m-PEG12-DBCO is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent features a DBCO group that enables strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing azide groups. PROTACs utilizing this linker connect an E3 ubiquitin ligase ligand with a target protein ligand to leverage the intracellular ubiquitin-proteasome system for selective target protein degradation.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-56625