Learn More
Medchemexpress LLC m-PEG12-Thiol | 2413368-61-1 | 95.0% | C25H52O12S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141331100MG
Encompass_Preferred
m-PEG12-Thiol is a PEG-based PROTAC linker, pivotal for synthesizing PROTACs. These bifunctional molecules are engineered to induce the degradation of specific target proteins within cells.
  • Composed of two distinct ligands connected by a linker
  • One ligand recruits an E3 ubiquitin ligase
  • The other ligand binds to the target protein for degradation
  • Utilizes the cell's ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-52953