Learn More
Medchemexpress LLC m-PEG12-Thiol | 2413368-61-1 | 95.0% | C25H52O12S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14133150MG
Encompass_Preferred
m-PEG12-Thiol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds that leverage the intracellular ubiquitin-proteasome system for the targeted degradation of proteins. These molecules consist of two distinct ligands, one binding to an E3 ubiquitin ligase and the other to a target protein, connected by a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates targeted protein degradation
Catalog No. 50-003-56781