Learn More
Medchemexpress LLC M-PEG13-NHS ester | 3063847-98-0 | 95.0% | C32H59NO17 | 50 MG
Supplier: Medchemexpress LLC HY14110750MG
M-PEG13-NHS ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
- Can be used in the synthesis of PROTACs
- PROTACs contain two different ligands connected by a linker
- One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest
- Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins