Learn More
Medchemexpress LLC M-PEG13-NHS ester | 3063847-98-0 | MFCD28385483 | 95.0% | 729.81 | C32H59NO17 | 5 MG

Supplier: Medchemexpress LLC HY1411075MG
m-PEG13-NHS ester is a PEG-based linker bearing an N-hydroxysuccinimide (NHS) ester reactive group, designed for conjugation to primary amines during the synthesis of proteolysis targeting chimeras (PROTACs). Intended for research use.
- Contains an NHS ester for efficient coupling to primary amines.
- Provides a PEG13 spacer to increase linker flexibility and solubility.
- Molecular weight 729.81; chemical formula C32H59NO17.
- Purity 95.0% and colorless to off-white oil appearance.
- Stable when sealed and protected from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
- Packaged in small quantities suitable for research-scale synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.