Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG15-NHS ester | 98.0% | 100 MG
SDP

Supplier:  Medchemexpress LLC HY130829100MG

Encompass_Preferred

m-PEG15-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. These Proteolysis Targeting Chimeras consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for a target protein. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This compound has a molecular weight of 875.99 and a chemical formula of C39H73NO20. It appears as a white to off-white solid below 35°C and a liquid above 35°C.

  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Molecular weight: 875.99
  • Chemical formula: C39H73NO20
  • Appears as a white to off-white solid below 35°C
  • Liquid above 35°C
  • Store at -20°C in sealed storage, away from moisture and light
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-003-40345


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.