Learn More
Medchemexpress LLC M-PEG16-COOH | 2933174-52-6 | 98.0% | 764.89 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13831450MG
Encompass_Preferred
m-PEG16-COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-50112