Learn More
Medchemexpress LLC m-PEG2-CH2CH2COOH | 209542-49-4 | MFCD20646047 | ≥97.0% | 192.21 g/mol | C8H16O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0437255G
Encompass_Preferred
m-PEG2-CH2CH2COOH is a PEG-based PROTAC linker used in the synthesis of proteolysis targeting chimeras (PROTACs). It provides a short, flexible polyethylene glycol spacer terminating in a carboxylic acid group for conjugation and linkage in medicinal chemistry research.
  • Short PEG2 spacer terminating in a carboxylic acid for conjugation.
  • Suitable for use as a PROTAC linker in medicinal chemistry applications.
  • Available in multiple sizes for synthesis scale flexibility, including 5 G.
  • Characterized by CAS 209542-49-4 and molecular weight 192.21 g/mol.
Catalog No. 50-002-10168