Learn More
Medchemexpress LLC m-PEG2-CH2CH2COOH | 209542-49-4 | MFCD20646047 | 97.0% | 192.21 g/mol | C8H16O5 | 500 MG

Supplier: Medchemexpress LLC HYW043725500MG
m-PEG2-CH2CH2COOH is a PEG2-based linker bearing a terminal carboxylic acid used as a spacer in the synthesis of PROTACs and for bioconjugation applications.
- Molecular formula C8H16O5 and molecular weight 192.21 g/mol.
- CAS number 209542-49-4.
- Typical purity 97.0% (NMR).
- Available in 250 mg, 500 mg, 1 g, 5 g, and larger sizes by quotation.
- Intended for use as a spacer/linker in medicinal chemistry and PROTAC synthesis.
- Store cold (examples indicate -20°C for long-term storage).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.