Learn More
Medchemexpress LLC M-PEG24-alcohol | 2376450-73-4 | 99.7% | C49H100O25 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141225100MG
Encompass_Preferred
M-PEG24-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Facilitates synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Applicable in cancer and cancer targeted therapy research
Catalog No. 50-003-56264