Learn More
Medchemexpress LLC m-PEG24-alcohol | 2376450-73-4 | 99.7% | C49H100O25 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY141225
Encompass_Preferred
m-PEG24-alcohol is a polyethylene glycol (PEG)-based PROTAC linker utilized in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • Stable under recommended storage conditions
  • Appears as an oil
  • Colorless to light yellow appearance
Catalog No. 50-003-52789