Learn More
Medchemexpress LLC M-PEG24-alcohol | 2376450-73-4 | 99.7% | C49H100O25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14122550MG
Encompass_Preferred
M-PEG24-alcohol is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are innovative molecules that consist of two distinct ligands joined by a linker. One ligand is engineered to bind to an E3 ubiquitin ligase, while the other targets a specific protein. This mechanism allows PROTACs to leverage the cell's natural ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based linker for PROTACs
  • Facilitates the synthesis of PROTACs
  • Enables targeted protein degradation via the ubiquitin-proteasome system
  • Connects a ligand for E3 ubiquitin ligase to a target protein ligand
Catalog No. 50-003-53828