Learn More
Medchemexpress LLC m-PEG24-NH2 | 2151823-08-2 | 99.9% | C49H101NO24 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1402285G
Encompass_Preferred
m-PEG24-NH2 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Exploits the intracellular ubiquitin-proteasome system
Catalog No. 50-003-57319