Learn More
Medchemexpress LLC M-PEG3-0-benzaldehyde | 153364-63-7 | 95% | C14H20O5 | 10 G
SDP
Supplier:  Medchemexpress LLC HY14062110G
Encompass_Preferred
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53512