Learn More
Medchemexpress LLC M-PEG3-0-benzaldehyde | 153364-63-7 | C14H20O5 | 100 MG

Supplier: Medchemexpress LLC HY140621100MG
m-PEG3-0-benzaldehyde is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. PROTACs work by utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
- PEG-based PROTAC linker
- Utilized in the synthesis of PROTACs
- Intended for research use only
- Molecular weight: 268.31
- Appearance: Liquid
- Color: Colorless to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.