Learn More
Medchemexpress LLC m-PEG3-Aminooxy | 248275-10-7 | 98.0% | C7H17NO4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14039850MG
Encompass_Preferred
m-PEG3-Aminooxy is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one binds to an E3 ubiquitin ligase and the other to the target protein. They function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs.
  • Acts as a PEG-based PROTAC linker.
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Intended for research use only.
Catalog No. 50-003-56640