Learn More
Medchemexpress LLC M-PEG3-azide | 74654-06-1 | 98.0% | C7H15N3O3 | 500 MG
Supplier: Medchemexpress LLC HY140827500MG
m-PEG3-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
- PEG-based PROTAC linker
- Click chemistry reagent
- Contains an Azide group
- Participates in CuAAc reactions with Alkyne groups
- Participates in SPAAC reactions with DBCO or BCN groups