Learn More
Medchemexpress LLC m-PEG4-C6-phosphonic acid ethyl ester | 2028281-89-0 | C19H41O7P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141314100MG
Encompass_Preferred
m-PEG4-C6-phosphonic acid ethyl ester is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. This compound, which appears as a colorless to light yellow liquid, has a molecular weight of 412.50. PROTACs are designed to degrade target proteins by exploiting the intracellular ubiquitin-proteasome system, typically featuring two ligands connected by a linker.
  • PEG-based PROTAC linker
  • Colorless to light yellow liquid appearance
  • Molecular weight: 412.50
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-56048