Learn More
Medchemexpress LLC m-PEG4-Hydrazide | 1449390-68-4 | >98% | C10H22N2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY141402100MG
Encompass_Preferred
M-PEG4-Hydrazide is a PEG-based PROTAC linker intended for research use only. It is utilized in the synthesis of PROTACs, which are Proteolysis-Targeting Chimeras. These chimeras consist of two ligands connected by a linker: one for an E3 ubiquitin ligase and another for the target protein. PROTACs employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Facilitates selective degradation of target proteins
  • Intended for research use only
Catalog No. 50-003-57071