Learn More
Medchemexpress LLC M-PEG4-hydrazide | 1449390-68-4 | >98% | C10H22N2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14140250MG
Encompass_Preferred
m-PEG4-Hydrazide is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs.
  • PROTACs are designed to contain two different ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest.
  • PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade specific target proteins.
Catalog No. 50-003-57070