Learn More
Medchemexpress LLC M-PEG4-PFP ester | 2952895-31-5 | ≥98.0% | 402.31 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13838250MG
Encompass_Preferred
m-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase, and the other for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-49300