Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG5-CH2CH2COOH | 1347750-72-4 | MFCD20926383 | >95.0% | 324.37 | C14H28O8 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0438405G

Encompass_Preferred

m-PEG5-CH2CH2COOH is a methyl-capped polyethylene glycol (PEG5) linker terminating in a carboxylic acid. It is used as a short PEG spacer for conjugation in PROTAC and ADC synthesis, providing solubility and a reactive terminal carboxyl for coupling reactions.

  • Methyl-capped PEG5 spacer with terminal carboxylic acid.
  • Designed for use in PROTAC and ADC linker synthesis.
  • Typical purity ≥95.0%.
  • Molecular weight 324.37 and formula C14H28O8.
  • Available in small-scale packages including 5 g.
  • Store pure material at -20°C for long-term stability.

Catalog No. 50-002-10221


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.