Learn More
Medchemexpress LLC m-PEG5-succinimidyl carbonate | 1058691-00-1 | MFCD30828680 | 98.0% | 393.39 g/mol | C16H27NO10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130150100MG
Encompass_Preferred
m-PEG5-succinimidyl carbonate is a non-cleavable, 5-unit polyethylene glycol (PEG) succinimidyl carbonate linker with an NHS-type carbonate reactive group for conjugation to primary amines. It is commonly used in synthesis of antibody-drug conjugates (ADCs), PROTAC linkers, and for PEGylation of amine-containing molecules. Typical specifications: purity 98.0%, molecular weight 393.39 g/mol, formula C16H27NO10, CAS 1058691-00-1.
  • Provides an NHS-type carbonate reactive group for efficient amine conjugation.
  • Non-cleavable 5-unit PEG spacer increases solubility and spacing between conjugated moieties.
  • High purity (98.0%) suitable for research-grade conjugation chemistry.
  • Available in small pack sizes to support synthetic workflows.
  • Stable when stored frozen; follow solvent storage recommendations for solutions.
Catalog No. 50-002-02143