Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG6-Br | 125562-29-0 | 99.8% | C13H27BrO6 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY141376

Encompass_Preferred

m-PEG6-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins, by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker.

  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Pure form storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-57349


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.