Learn More
Medchemexpress LLC m-PEG6-CH2CH2COOH | 874208-91-0 | MFCD20926396 | ≥95.0% | 368.42 g·mol⁻¹ | C16H32O9 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW040239
Encompass_Preferred
m-PEG6-CH2CH2COOH is a polyethylene glycol (PEG)-based linker intended for use as a chemical building block in the synthesis of PROTACs and other bioconjugates. It is provided as a research reagent with high purity and defined molecular properties suitable for medicinal chemistry and linker optimization workflows.
  • PEG-based linker suitable for PROTAC and bioconjugation applications.
  • High purity (≥95%) supports reliable reaction performance.
  • Defined molecular weight (368.42 g·mol⁻¹) for stoichiometric calculations.
  • Supplied in small pack sizes convenient for medicinal chemistry research.
  • Available CAS identifier for unambiguous chemical reference.
Catalog No. 50-002-09474