Learn More
Medchemexpress LLC M-PEG6-Tos | 155887-96-0 | 99.6% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130336
Encompass_Preferred
M-PEG6-Tos is a highly pure PEG-based PROTAC linker, with a purity of 99.6%. It is specifically designed for the synthesis of PROTACs, molecules that harness the body's natural ubiquitin-proteasome system to selectively degrade target proteins. This linker plays a crucial role in forming PROTACs by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein, enabling targeted protein degradation.
  • PEG-based PROTAC linker.
  • Used for synthesizing PROTACs.
  • Facilitates targeted protein degradation.
  • Connects E3 ubiquitin ligase ligand to target protein ligand.
  • High purity of 99.6%.
Catalog No. 50-003-41490