Learn More
Medchemexpress LLC m-PEG7-alcohol | 4437-01-8 | 98.6% | C15H32O8 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY141217
Encompass_Preferred
m-PEG7-alcohol is a PEG-based PROTAC linker for use in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52820