Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG7-alcohol (O-Methyl-heptaethylene glycol) | 4437-01-8 | 98.56% | C15H32O8 | 100 G
SDP

Supplier:  Medchemexpress LLC HY141217100G

Encompass_Preferred

m-PEG7-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity: 98.56%
  • Molecular weight: 340.41
  • Chemical formula: C15H32O8
  • CAS number: 4437-01-8
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Supports the development of targeted therapy drugs for cancer
  • Utilizes the intracellular ubiquitin-proteasome system for protein degradation

Catalog No. 50-003-53825


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.