Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG7-alcohol (O-Methyl-heptaethylene glycol) | 4437-01-8 | 98.56% | C15H32O8 | 5 G
SDP

Supplier:  Medchemexpress LLC HY1412175G

Encompass_Preferred

m-PEG7-alcohol (O-Methyl-heptaethylene glycol) is a PEG-based PROTAC linker. It is for research use only and can be used in the synthesis of PROTACs.

  • Purity: 98.56%
  • Appearance: Colorless to light yellow liquid
  • Molecular Weight: 340.41
  • Density: 1.069±0.06 g/cm3

Catalog No. 50-003-56022


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.