Learn More
Zymo Research Corporation M-Solubilization Buffer, 18 ml
SDP
Supplier:  Zymo Research Corporation D50217
Encompass_Preferred
The M-Solubilization Buffer facilitates the solubilization of the CT Conversion Reagent from a solid mixture into a liquid reagent. Highlights: Sold separately or as part of the EZ DNA Methylation-Direct Kits. Storage temperature: room temperature.
Catalog No. 50-444-329