Learn More
Cytiva M-TGE Broth with TTC Indicator, Total Bacteria , 2 mL plastic (50/pkg)
Supplier:  Cytiva 68111
Encompass
Sterile Ampoule Media Pre-Measured for Maximum Efficiency and Convenience-Meets Membrane Filter (MF) Technique Standard Method requirements with 2 mL ampoules*-Maximizes efficiency with premixed and presterilized media-Choose between plastic or glass ampoules for your applications-Simplify pouring of the media with wide-mouth glass ampoules-All plastic ampoule media is economically packaged with 50 ampoules per box This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3358