Learn More
Teledyne ISCO, Inc M1 Class Single Piston Pump; Eccentric Drive; 0.00 - 10.00 Ml/Min.; 2,000 Psi
Supplier:  Teledyne ISCO, Inc M1010SRX
Encompass
M1 Class Single Piston Pump; Eccentric Drive; 0.00 - 10.00 mL/min.; 2,000 psi; Stainless Steel Fluid Path; Automatic Piston Wash; without Pulse Dampener; Standard Seal (stainless steel energizer);Fitting package for 1/8"""" and 1/16"""" tubing
Catalog No. 50-211-9952