Learn More
Sino Biological Anti-M13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 68007T48100
Encompass_Preferred
Produced in rabbits immunized with purified M13 Bacteriophage coat protein G3P (YP_009111293.1, Ala19-Ala130). phage coat protein g3p of fd phage or M13 phage specific IgG was purified by M13 Bacteriophage coat protein G3P (P69168, Ala19-Ala130) affinity chromatography.
Catalog No. 50-222-1613