Learn More
Medchemexpress LLC M199 100mg | 1051933-86-8 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY114592100MG
Encompass_Preferred
M199 is a small-molecule inhibitor of Toll-like receptor 3 (TLR3) and Toll-like receptor 9 (TLR9) signaling that induces secretion of interleukin-6 (IL-6), interleukin-8 (IL-8), and tumor necrosis factor α (TNFα) in human peripheral blood mononuclear cells. It is used as a selective inducer/modulator of immune responses in research settings.
  • Inhibits TLR3 and TLR9 signaling pathways
  • Induces IL-6, IL-8, and TNFα secretion in human PBMCs
  • Useful for studying innate immune activation and cytokine responses
  • High chemical purity for reproducible experimental results
  • Available in small research quantities including 100 mg
Catalog No. 50-002-29545