Learn More
Medchemexpress LLC M199 5mg | 1051933-86-8 | 5 MG

Supplier: Medchemexpress LLC HY1145925MG
M199 is a small-molecule inhibitor of Toll-like receptor 3 and 9 (TLR3/TLR9) used in research to modulate innate immune signaling. It induces secretion of IL-6, IL-8, and TNFα in human peripheral blood mononuclear cells (PBMCs) and is supplied as a solid for laboratory use.
- Potent inhibitor of TLR3 and TLR9 signaling.
- Induces secretion of IL-6, IL-8, and TNFα in human PBMCs.
- Molecular formula: C17H17N3O; molecular weight: 279.34.
- Purity: 98.6% (as listed by the supplier).
- Appearance: solid, yellow to brown.
- Storage: powder at -20°C for long-term; in solvent, store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.