Learn More
Medchemexpress LLC M2n12 | 2376577-06-7 | 98.2% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1287691MG
Encompass_Preferred
M2N12 is a potent and highly selective cell division cycle 25C protein phosphatase (Cdc25C) inhibitor. It also shows promising activity against Cdc25A and Cdc25B. M2N12 exhibits anti-tumor activity and is suitable for cancer research.
  • Potent and highly selective Cdc25C inhibitor with an IC50 value of 0.09 μM.
  • Active against Cdc25A (IC50: 0.53 μM) and Cdc25B (IC50: 1.39 μM).
  • Exhibits anti-tumor activity, suitable for cancer research.
  • Demonstrates cytotoxic activity against multiple tumor cell lines, including A549, KB, and MCF7.
  • Appearance is solid, brown to red.
Catalog No. 50-003-40550