Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12876910MM/1ML
Encompass_Preferred
M2N12 is a potent and highly selective inhibitor of cell division cycle 25C protein phosphatase (Cdc25C) with an IC50 value of 0.09 μM. It also shows activity against Cdc25A (IC50 = 0.53 μM) and Cdc25B (IC50 = 1.39 μM). This product exhibits anti-tumor activity and is suitable for cancer research.
  • Potent and highly selective inhibitor of Cdc25C.
  • Shows activity against Cdc25A and Cdc25B.
  • Exhibits anti-tumor activity.
  • Suitable for cancer research.
  • Exhibits cytotoxic activity against several tumor cell lines.
Catalog No. 50-003-39605