Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12876910MG
Encompass_Preferred
M2N12 is a potent and highly selective inhibitor of cell division cycle 25C protein phosphatase (Cdc25C) with an IC50 value of 0.09 μM. It also shows promising activity against Cdc25A and Cdc25B, with IC50 values of 0.53 μM and 1.39 μM, respectively. M2N12 exhibits anti-tumor activity and is suitable for cancer research. It has cytotoxic activity against several tumor cell lines, including A549, MDA-MB-231, KB, KB-VIN, and MCF-7, as well as a normal cell line (HBE).
  • Potent and selective Cdc25C inhibitor
  • Active against Cdc25A and Cdc25B
  • Exhibits anti-tumor activity
  • Suitable for cancer research
  • Cytotoxic activity against multiple tumor cell lines
  • Cytotoxic activity against normal cell line HBE
Catalog No. 50-003-42850