Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128769100MG
Encompass_Preferred
M2N12 is a potent and highly selective cell division cycle 25C protein phosphatase (Cdc25C) inhibitor with an IC50 value of 0.09 μM. It also shows promising activity against Cdc25A and Cdc25B with IC50 values of 0.53 μM and 1.39 μM, respectively. M2N12 exhibits anti-tumor activity and can be utilized for cancer research.
  • Potent and highly selective Cdc25C inhibitor
  • Active against Cdc25A and Cdc25B
  • Exhibits anti-tumor activity
  • Suitable for cancer research
Catalog No. 50-003-41951