Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128769
Encompass_Preferred
M2N12 is a potent and highly selective Cdc25C protein phosphatase inhibitor with an IC50 of 0.09 μM. It also demonstrates promising activity against Cdc25A and Cdc25B, with IC50 values of 0.53 μM and 1.39 μM, respectively. This compound exhibits anti-tumor activity and is suitable for cancer research.
  • Potent and highly selective cell division cycle 25C protein phosphatase (Cdc25C) inhibitor
  • IC50 value against Cdc25C is 0.09 μM
  • Shows activity against Cdc25A (IC50: 0.53 μM) and Cdc25B (IC50: 1.39 μM)
  • Possesses anti-tumor activity
  • Suitable for cancer research
  • Cytotoxic activity against various tumor cell lines (A-549, MDA-MB-231, KB, KB-VIN, MCF-7) and a normal cell line (HBE)
Catalog No. 50-003-43074