Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 393.83 | 5 MG

Supplier: Medchemexpress LLC 5MGHY128769
M2N12 is a potent and highly selective Cdc25C protein phosphatase inhibitor. It demonstrates activity against Cdc25A and Cdc25B, and possesses anti-tumor properties, making it suitable for cancer research.
- Potent and highly selective Cdc25C inhibitor (IC50 = 0.09 μM)
- Active against Cdc25A (IC50 = 0.53 μM) and Cdc25B (IC50 = 1.39 μM)
- Exhibits anti-tumor activity
- Suitable for cancer research applications
- Appears as a brown to red solid
- Soluble in DMSO (11.11 mg/mL)
- Effective against multiple tumor cell lines including A549, KB, and MCF7
- Store powder at -20°C for 3 years, or 4°C for 2 years
- Store in solvent at -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.