Learn More
Medchemexpress LLC M2N12 | 2376577-06-7 | 98.2% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12876950MG
Encompass_Preferred
M2N12 is a potent and highly selective cell division cycle 25C protein phosphatase (Cdc25C) inhibitor with an IC50 value of 0.09 μM. It also shows activity against Cdc25A and Cdc25B with IC50 values of 0.53 μM and 1.39 μM, respectively. This compound possesses anti-tumor activity and is suitable for cancer research.
  • Potent and highly selective Cdc25C inhibitor.
  • Active against Cdc25A and Cdc25B.
  • Demonstrates anti-tumor activity.
  • Can be used for cancer research.
  • Exhibits cytotoxic activity against multiple tumor cell lines (A-549, MDA-MB-231, KB, KB-VIN, MCF-7) and a normal cell line (HBE).
Catalog No. 50-003-39407