Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M4 mAChR agonist-1 | 785705-53-5 | 98.95% | 290.38 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY14702850MG

Encompass_Preferred

M4 mAChR agonist-1 is a potent M4 mAChR agonist with an EC50 >10 μM for human M4 receptors. This solid, light yellow to brown substance has the chemical formula C14H18N4OS and is intended for research use only.

  • Potent M4 mAChR agonist
  • EC50 >10 μM for human M4
  • Solid, light yellow to brown appearance
  • Purity of 98.95%
  • Molecular weight of 290.38
  • For research use only
  • Store at 4°C, protected from light
  • Soluble in DMSO

Catalog No. 50-003-62400


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.