Learn More
ATCC M4A4 GFP Human Metastatic Breast Cancer Cell Line, GFP-expressing
Supplier:  ATCC CRL2915
Encompass
M4A4 GFP is a human, tumorigenic cell line engineered to express green fluorescent protein (GFP). Developed by transducing GFP into the parental M4A4 line and is part of an isogenic set exhibiting contrasting metastatic behavior, making it suitable for metastatic studies, assay development, and drug screening.
  • Derived from a human breast cancer background and engineered to express GFP for fluorescence-based assays.
  • Well characterized tumorigenic line with a highly metastatic phenotype in immunodeficient mice.
  • Suitable for metastatic studies, assay development, drug development, and high-throughput screening.
  • Grows with epithelial morphology and is maintained in Dulbecco's Modified Eagle's Medium with 10% fetal bovine serum.
  • Maintain at 37°C in 95% air, 5% CO2; store in the vapor phase of liquid nitrogen for long-term preservation.
  • Compatible with 3D cell culture and imaging applications that leverage GFP expression.
Catalog No. NC2938817