Learn More
IDEX Health & Science Upchurch M6 FEMALE TO MALE LUER
Supplier:  IDEX Health & Science Upchurch P657
Encompass_Preferred
Idex Threaded Luer PEEK Adapter, Black, For Use With: To Female M6x1, Luer Adapter, Diameter Inner: 0.05 in., Temperature Max: 125 deg.C
Catalog No. NC1919473