Learn More
Diapharma Group, Inc. M65 EpiDeath CK18 Kit, 96 tests
SDP
Supplier:  Diapharma Group, Inc. P10040
Encompass
The M65 EpiDeath ELISA kit measures the concentration of soluble CK18 (K18) in cell culture supernatants, serum or plasma for research studies. CK18 levels reflect the amount of total epithelial cell death, regardless of the cause of death.
Catalog No. NC0266302