Learn More
Medchemexpress LLC M65 TFA | 99.98% | 4823.53 (free base) | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP4127A1MG
Encompass_Preferred
M65 TFA is a deleted peptide of maxadilan (61 a.a.) with a deletion of residues between positions 24 and 42. It acts as a specific antagonist of the PACAP type 1 receptor, inhibiting ANP secretion, and can be utilized for relevant research purposes.
  • Specific antagonist of the PACAP type 1 receptor.
  • Inhibits ANP secretion.
  • Suitable for research.
  • In vitro activity: M65 (1 μM) completely blocks cAMP accumulation stimulated by 100 nM of VIP and partially inhibits cAMP accumulation stimulated by 1 nM of maxadilan in rat cortical neurons.
Catalog No. 50-003-99382