Learn More
Medchemexpress LLC m7GpppAmpG ammonium | 62858-30-4 | 99.5% | C32H43N15O24P4.xNH3 | 20 UL
SDP
Supplier:  Medchemexpress LLC HY145974A1.1MG
Encompass_Preferred
m7GpppAmpG ammonium is a trinucleotide 5′ end cap analog that binds to eIF4E and caps RNA. It enhances mRNA stability and translation efficiency, making it valuable for mRNA therapeutic research.
  • Trinucleotide 5′ end cap analog
  • Binds to eIF4E with a KD value of 45.6 nM
  • Caps RNA with 90% efficiency
  • Enhances mRNA stability
  • Enhances translation efficiency
  • Used in mRNA therapeutic research
Catalog No. 50-003-55664